missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, rutile, single crystal, Thermo Scientific™

Product Code. 15491447
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491447 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491447 Supplier Thermo Scientific Alfa Aesar Supplier No. 036425.KT
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1317-80-2
Quantity 1 Ea.
Molecular Formula O2Ti
MDL Number MFCD00011269,MFCD00210650
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES O=[Ti]=O
IUPAC Name dioxotitanium
Molecular Weight (g/mol) 79.87
PubChem CID 26042
ChEBI CHEBI:32234
Formula Weight 79.9
Odor Odorless
Density 4.23 g/mL
Chemical Name or Material Titanium(IV) oxide, rutile, Single crystal, 0.5mm thick x 10mm x 10mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.