missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Low Alloyed, Grade 12, R53400, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Low Alloyed, Grade 12, R53400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Hydrogen/Copper/Tin/Nitrogen/Vanadium/Chromium/Carbon/Silicon/Aluminium/Iron/Oxygen/Molybdenum/Nickel/Cobalt/Magnesium/Phosphorus/Hafnium/Tantalum/Ruthenium/Tungsten/Manganese/Yttrium/Boron/Palladium/Niobium/Titanium |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0016 %/Hydrogen 0.0027 %/Copper 0.003 %/Tin 0.004 %/Nitrogen 0.006 %/Vanadium 0.006 %/Chromium 0.008 %/Carbon 0.012 %/Silicon 0.012 %/Aluminium 0.013 %/Iron 0.095 %/Oxygen 0.15 %/Molybdenum 0.288 %/Nickel 0.88 %/Cobalt (<0.002) %/Magnesium (<0.002) %/Phosphorus (<0.003) %/Hafnium (<0.004) %/Tantalum (<0.005) %/Ruthenium (<0.006) %/Tungsten (<0.03) %/Manganese (<0.05) %/Yttrium (0.0004) %/Boron (0.0005) %/Palladium (0.002) %/Niobium (0.004) %/Titanium (98.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction