missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.5% (metals basis)

Product Code. 15454845
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454845 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15454845 Supplier Thermo Scientific Alfa Aesar Supplier No. 041111.KS
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25583-20-4 | NTi | 61.87 g/mol

Titanium nitride is used in coating on titanium alloys, steel, carbide, and aluminum components to improve the substrate's surface properties. Used to harden and protect cutting and sliding surfaces, for decorative purposes. It is used to coat costume jewelry and automotive trim. It is also used in also used in microelectronics. It is also used in top layer coating, usually with nickel to chromium plated substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Quantity 1 Ea.
CAS 25583-20-4
Form Hot Pressed Material
Molecular Formula NTi
MDL Number MFCD00049596
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
Solubility Information Soluble in aqua regain. Insoluble in water.
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILES N#[Ti]
IUPAC Name azanylidynetitanium
Molecular Weight (g/mol) 61.87
PubChem CID 93091
Formula Weight 61.91
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.