missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis), Thermo Scientific™

Product Code. 15464845
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15464845

Brand: Thermo Scientific Alfa Aesar 041112.KS

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 2950°C
CAS 25583-20-4
Color Gray
Physical Form Hot Pressed Material
Quantity 1 Ea.
Molecular Formula NTi
MDL Number MFCD00049596
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILES N#[Ti]
IUPAC Name azanylidynetitanium
Molecular Weight (g/mol) 61.87
PubChem CID 93091
Formula Weight 61.91
Percent Purity 99.5%
Density 5.22 g/mL
Assay (metals basis)
Chemical Name or Material Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.