missing translation for 'onlineSavingsMsg'
Learn More

Titanium plate, 6mm (0.2in) thick, 100x100mm (3.9x3.9in), Puratronic™, 99.999% (metals basis)

Product Code. 15444765
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444765 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444765 Supplier Thermo Scientific Alfa Aesar Supplier No. 036680.KG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-32-6
Quantity 1 Pc.
Odor Odorless
Weight ≈271g/pc
Form Plate
Molecular Formula Ti
MDL Number MFCD00011264,MFCD00151301
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES [Ti]
IUPAC Name titanium
Molecular Weight (g/mol) 47.87
PubChem CID 23963
ChEBI CHEBI:33341
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Titanium plate, 6mm (0.2 in.) thick, 100x100mm (3.9 x 3.9 in.)
Show More Show Less

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.