missing translation for 'onlineSavingsMsg'
Learn More

Titanium Tablets, BUCHI

Product Code. 13564700
Click to view available options
Quantity:
1000 Tablets
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Quantity 1000 Tablets
Composition 3.5g K2SO4 / 0.105g CuSO4 • 5 H2O 0.105g TiO2
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Kjeldahl Tablets Titanium
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : copper(II) sulphatepentahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'