missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 3-2.5, Grade 9, R56320, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 3-2.5, Grade 9, R56320, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Hydrogen/Copper/Zirconium/Molybdenum/Nitrogen/Tin/Silicon/Carbon/Chromium/Nickel/Oxygen/Iron/Vanadium/Aluminium/Yttrium/Ruthenium/Tungsten/Palladium/Manganese/Niobium/Phosphorus/Cobalt/Titanium |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0004 %/Hydrogen 0.001 %/Copper 0.003 %/Zirconium 0.003 %/Molybdenum 0.004 %/Nitrogen 0.005 %/Tin 0.006 %/Silicon 0.007 %/Carbon 0.011 %/Chromium 0.014 %/Nickel 0.016 %/Oxygen 0.085 %/Iron 0.18 %/Vanadium 2.46 %/Aluminium 3.05 %/Yttrium (0.0004) %/Ruthenium (0.001) %/Tungsten (0.001) %/Palladium (0.002) %/Manganese (0.003) %/Niobium (0.003) %/Phosphorus (0.003) %/Cobalt (0.005) %/Titanium (94.1) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction