missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 6-7, R56700, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 6-7, R56700, Ø32x3mm
Specifications
Specifications
| Chemical Name or Material | Molybdenum/Hydrogen/Nitrogen/Tin/Vanadium/Tantalum/Manganese/Carbon/Nickel/Zirconium/Copper/Silicon/Chromium/Oxygen/Iron/Aluminium/Niobium/Magnesium/Phosphorus/Tungsten/Yttrium/Boron/Cobalt/Palladium/Titanium |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Molybdenum 0.0013 %/Hydrogen 0.0025 %/Nitrogen 0.003 %/Tin 0.004 %/Vanadium 0.004 %/Tantalum 0.005 %/Manganese 0.006 %/Carbon 0.0081 %/Nickel 0.012 %/Zirconium 0.013 %/Copper 0.014 %/Silicon 0.016 %/Chromium 0.0174 %/Oxygen 0.171 %/Iron 0.175 %/Aluminium 6.09 %/Niobium 6.84 %/Magnesium (<0.002) %/Phosphorus (<0.004) %/Tungsten (<0.005) %/Yttrium (<0.005) %/Boron (0.0012) %/Cobalt (0.002) %/Palladium (0.005) %/Titanium (86.3) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction