missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 6-7, R56700, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 6-7, R56700, Ø35x19mm
Specifications
Specifications
| Chemical Name or Material | Molybdenum/Copper/Nitrogen/Chromium/Carbon/Nickel/Silicon/Vanadium/Oxygen/Iron/Aluminium/Niobium/Lead/Magnesium/Phosphorus/Tungsten/Palladium/Antimony/Boron/Hydrogen/Cobalt/Yttrium/Manganese/Tantalum/Zirconium/Tin/Titanium |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Molybdenum 0.0023 %/Copper 0.003 %/Nitrogen 0.0061 %/Chromium 0.008 %/Carbon 0.01 %/Nickel 0.014 %/Silicon 0.018 %/Vanadium 0.03 %/Oxygen 0.174 %/Iron 0.181 %/Aluminium 5.97 %/Niobium 6.8 %/Lead (<0.0001) %/Magnesium (<0.001) %/Phosphorus (<0.001) %/Tungsten (<0.002) %/Palladium (<0.005) %/Antimony (0.0005) %/Boron (0.0006) %/Hydrogen (0.0006) %/Cobalt (0.001) %/Yttrium (0.002) %/Manganese (0.003) %/Tantalum (0.005) %/Zirconium (0.005) %/Tin (0.006) %/Titanium (86.7) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction