Learn More
BRAND™ Titrating Tube for Titrette™ Bottletop Burette
Product Code. 10798195 Shop All Brand Products
Click to view available options
For Use With (Equipment):
10 mL Titrette
25 mL Titrette
Unit Size:
Each
Product Code. 10798195
Brand: BRAND™ 707526
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes integrated discharge/recirculation valve.
  • For use with Titrette™ Bottle Top Burette
  • Screw cap fits onto titrating tube to prevent leakage
  • Includes discharge/recirculation valve
TRUSTED_SUSTAINABILITY
Specifications
Product Type Titrating Tube
For Use With (Equipment) 25 mL Titrette
Includes Screw cap for tube, discharge/recirculation tube
Quantity 1 Ea.