missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Titrator TitroLine™ 7500 KF Trace Module 3

Product Code. 15372477 Shop All SI Analytics Products
Change view
Click to view available options
Frequency:
50/60 Hz
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frequency Unit Size
15372477 50/60 Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15372477 Supplier SI Analytics™ Supplier No. 285220880
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TitroLine™ 7500 KF trace is a compact, easy to use coulometric titrator for water determination according to Karl Fischer (KF) with a typical operating range of 1 ppm - 5% water.

  • Automatic titrator for the coulometric KF-Titration
  • High visibility, full color display that can be easily viewed from a distance and at extreme angles
  • Includes up to three USB, one LAN and two RS232 ports for expansion and connection of devices
  • Export the results as PDF or CSV, also to networks
  • Specific features for the TitroLine 7500 KF trace
  • Coulometric KF titration with automatic drift determination and correction
  • With standard methods for various applications such as water in %, water in ppm, water with KF oven, etc.
  • Statistical functions average value and relative standard deviation
  • Configurable output: Short, standard with curve, GLP
  • Output of results in ppm, %, mg, μg, mg/L. Unit individually adjustable
  • Also suitable for the determination of bromine number
  • Formula editor for individual calculations
  • User management/password
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC compatibility according to the Council Directive: 2004/108/EG; applied harmonized standards: EN 61326-1:2006; Low-voltage directive according to the Council Directive 2006/95/EG Testing basis EN 61 010, Part 1
Data Management Details 3 x USB (2 x USB-A and 1 x USB-B), 2 x RS-232-C and 1 x LAN
Includes Basic unit, generator electrode TZ 1752 with junction + connection cable, magnetic stirrer TM 235 (up to 500 mL, 500 to 2000 rpm), stand rod, titration vessel TZ 1751, micro double platinum electrode KF 1150, power supply
Material Polypropylene Housing, Polyester Coated Front Keyboard
Measurement Modes Measuring input indicator electrode: Connector for double platinum electrode with 2 x 4 mm sockets
Power Supply 100 to 240 V; 50/60 Hz, power input: 30 VA
Relative Humidity Range Max. 80% for temperatures up to 31°C, linear decrease down to 50% relative humidity at a temperature of 40°C
Temperature Range (Metric) Ambient 10°C to 40°C
Voltage 100/240 V; power input: 30 VA
Frequency 50/60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.