Learn More
BRAND™ Titrette™ Adapter Set Interface RS232 to USB
Product Code. 17821252 Shop All Brand Products
Change view
Click to view available options
For Use With (Application):
Connecting Burette to Computer
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
17821252 Connecting Burette to Computer Each
1 options
Product Code. 17821252 Supplier BRAND™ Supplier No. 707539
  • ONLINE EXCLUSIVE PRICE
    234.00 €
    202.36 € / Pack of 1
    Save 31.64 €
    13% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use in connecting the Titrette burette to a computer.
  • Adapter connects the Titrette™ bottle-top burette (with RS232 interface) to a desktop computer or laptop
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 17.6 cm
For Use With (Equipment) Computer or Laptop
For Use With (Application) Connecting Burette to Computer
Height (Metric) 3.23 cm
Includes RS232 Interface
Type USB Adapter