missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Beinhaltet
Cat. No. 15314408 - TitroLine 5000 with 20mL dosing unit
Ready to use assembled 20mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V
Cat. No. 15324408 - TitroLine 5000 with 50mL dosing unit
pH electrode and buffer set, with ready to use assembled 50mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V
- High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
- pH/mV-input with electrode socket according to DIN 19 262 or additional with BNC socket insert (Z 860)
- Pt 1000 and NTC 30 temperature measurement input (socket 2 x 4mm) for automatic temperature compensation
- Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
- Unit operated using the keys on the device itself, the controller TZ 3880 and optional PC-keyboard (USB)
- Graphics-capable TFT display
- Linear and dynamic titration to equivalence points
- Titration to pH and mV-ends points
- Stirrer Connection: TM 50 power supply directly through piston burette
- High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
- Same manual titration and dosing function as the piston burette Titronic™ 300
- Typical applications:
- pH-value, alkalinity (p+m-value)
- FOS/TAC
- Total Kjeldahl nitrogen
- Permanganate index and COD
- Chloride in wastewater
Zertifizierungen
CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95
Compliance
ISO 8655, part 6
Spezifikation
Spezifikation
| Certifications/Compliance | ISO 8655, part 6, CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95 |
| Connector Types | 1 x USB-A and 1 x USB-B, 2 x RS-232-C |
| Description | Automatic Titrator |
| Display Type | Graphics-capable TFT |
| Type | Titrator with Electrode |
| For Use With (Application) | Determine chloride in wastewater, permanganate index and COD, total Kjeldhal nitrogen, FOS/TAC, pH value, alkalinity |
| Includes | Basic unit with pH electrode and buffer set, with ready to use assembled 20mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100-240V |
| Model | Titrator, 20 mL Dosing Unit |
| Resolution | Display resolution: 0.005 to 0.025mL; Burette resolution: 8000 |
| Temperature Range (Metric) Ambient | 10°C to 40°C |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?