Description
- Intelligent exchangeable units store all relevant reagent and unit data
- Automatic wireless recognition of Schott Instruments ID electrodes guarantees accurate calibration and measurements
- Touch-sensitive glass keypad
- Three USB and two RS-232 ports
- High resolution pH/mV electrode and temperature inputs for pH, ISE, redox or photometric titrations
- Polarizable electrode input for set endpoint applications
- Available standard methods such as FOS/TAC, alkalinity, total acidity in soft drinks
- Linear (fixed increment) and dynamic equivalence point titration modes
- Titrations to pH, mV and μA end point
- Manual titration mode and routine dosing tasks are also available
Specifications
Specifications
| Connector Types | Three USB and two RS-232 ports |
| Description | Titrator |
| Display Type | TFT |
| For Use With (Application) | Food, water, wastewater and environmental applications |
| Includes | TitroLine 6000 basic unit, magnetic stirrer, 20mL exchangeable unit, bottle, adapters, hoses, drip off tube, titration tip; with pH combination electrode and buffer set |