missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 7500 KF trace M1 Titrator

Product Code. 15146052 Shop All SI Analytics Products
Change view
Click to view available options
Type:
Coulometric KF-Titrator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15146052 Coulometric KF-Titrator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15146052 Supplier SI Analytics™ Supplier No. 285220860
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase precision with high visibility full color display and standard methods for different applications. SI Analytics™ TitroLine™ 7500 KF trace M1 Titrator possesses general titration feautres but offers improved performace.

  • Full color display can be easily viewed from a distance and extreme angles
  • Store results via USB port (PDF - and CSV - format)
  • Includes standard methods for different applications (titer, blank value, 1 or 2 component reagent)
  • Titration stand manages the addition of solvent and titrated sample extraction
  • Accuracy: 0.3% at 1mg water
  • Includes: Coulometric KF-titrator, scope of supply: titrator unit, generator electrode TZ 1752 without diaphragm, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1754, micro-dual platinum electrode KF 11
TRUSTED_SUSTAINABILITY

Specifications

Type Coulometric KF-Titrator
Model TL 7500 KF trace M1
Temperature Range (Metric) Ambient 10°C to 40°C
Width (Metric) 15.3 cm
Depth (Metric) 29.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.