Learn More
SI Analytics™ TitroLine™ 7750
Product Code. 15524923 Shop All SI Analytics Products
Change view
Click to view available options
Model:
TitroLine 7750
TitroLine 7750 KF Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15524923 TitroLine 7750 KF Kit Each
13496768 TitroLine 7750 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15524923 Supplier SI Analytics™ Supplier No. 285220950
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SI Analytics™ TitroLine™ 7750 is the all-rounder for both potentiometric titration and volumetric KF titration.
  • pH/mV-input with 24 bit transducer Electrode socket according to DIN 19 262 or additional with BNC socket insert RFID receiver for SI Analytics ID electrodes
  • Connector (μA) for double platinum electrodes Polarisation voltage adjustable from 40 to 220 mV
  • Highly visible full color display, that can be easily viewed from a distance and extreme angles With new interchangeable modules which all relevant reagent and unit data can be stored
  • Expandable thanks to the 2 x USB-host, 1 x USB-PC, 1 x LAN and 2 x RS232 ports
  • Connectable are e.g. USB keyboard, USB printer, barcode reader, USB flash drives, balances, PC and further devices such as piston burettes and sample changers
  • Storage of results via USB or LAN connection, including method transfer
TRUSTED_SUSTAINABILITY
Specifications
Includes KF accessories: basic titrator unit, exchange unit WA 20, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1770, micro double platinum electrode KF 1100 and starter kit
Material Polypropylene
Model TitroLine 7750 KF Kit
Range (pH) -3 to 18
Temperature Range (Metric) Ambient 10°C to 40°C