Learn More
TUTTNAUER Tiva 15 Washer Disinfector with 2 Automatic Sliding Door
Product Code. 30058120 Shop All Tuttnauer Products
Change view
Click to view available options
Voltage:
380/400 V
Unit Size:
1 unit
Product Code. Voltage unitSize
30058120 380/400 V 1 unit
1 options
Product Code. 30058120 Supplier TUTTNAUER Supplier No. TIVA152V
Provides a flexible turnkey solution for washing, disinfecting and drying diverse medical instruments.
  • Equipped with automatic sliding door/s and is best suited for surgical and sterilization departments.
  • It can be loaded with up to 6 washing levels for a total 18 DIN baskets with a large variety of washing trolleys and accessories.
Specifications
Display LCD
Material Stainless Steel
Product Type Glassware Washer with Drying System and Steam Condenser
Style Dual Automatic Sliding Doors
Voltage 380/400 V