missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ TLS2200™ Portable Printer Accessories
TLS2200™ Portable Printer Accessories are for use with Brady TLS2200 Portable Printer (mfr. no. 19150079, sold separately).
22.70 € - 96.05 €
Specifications
Color | Black |
---|---|
For Use With (Equipment) | TLS 2200 Portable Thermal Transfer Printer |
Product Code | Brand | Product Type | Certifications/Compliance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Certifications/Compliance | Price | Quantity & Availability | ||||
10009891
|
Brady™
018556 |
Cleaning Kit | RoHS |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13085823
|
Brady™
218558 |
Battery Charger | RoHS, UL, CUL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Printer cleaning kits provide an easy solution for keeping your printer in top condition, For use in cleaning the TLS 2200™ and BMP™61 Label Printers, includes printer cleaning cards and swabs
- Printer Battery - can be run while battery is charging
- AC Adaptor - for use with BMP™71, TLS 2200™ and HandiMark™ Label Printers, power supply formatted for North America
Certifications
FCC Class A approved.
Specifications
Black | |
TLS 2200 Portable Thermal Transfer Printer |