missing translation for 'onlineSavingsMsg'
Learn More

TM-25659, MedChemExpress

Product Code. p-200099842
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487447

Brand: Medchem Express HY112920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TM-25659 is a transcriptional co-activator with PDZ-binding motif (TAZ) modulator. Anti-osteoporotic and anti-obesity activities.

TM-25659 is a transcriptional co-activator with PDZ-binding motif (TAZ) modulator. Anti-osteoporotic and anti-obesity activities.

Specifications

Chemical Name or Material TM-25659
CAS 260553-97-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H28N8
Quantity 5 mg
Solubility Information DMSO : ≥ 135 mg/mL (269.68 mM)
SMILES CC1=C(C2=CC=CN=C2)C=C3C(N(CC4=CC=C(C5=CC=CC=C5C6=NN=NN6)C=C4)C(CCCC)=N3)=N1
Molecular Weight (g/mol) 500.6
Formula Weight 500.6
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.