missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress TMB dihydrochloride

Product Code. 16499457
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499457 5 g 5g
16401589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499457 Supplier MedChemExpress Supplier No. HY15930A
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TMB dihydrochloride (BM blue dihydrochloride) is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualising reagent used in enzyme-linked immunosorbent assays (ELISA).

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 5 g
Shipping Condition Room temperature
Synonym BM blue dihydrochloride ∣ Sure Blue TMB dihydrochloride
CAS 64285-73-0
Molecular Weight (g/mol) 313.27 g/mol
Solubility DMSO : ≥ 33 mg/mL (105.34 mM) ∣ H2O : 11.11 mg/mL (35.46 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.