missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMB ELISA Buffer Kit, PeproTech®

Product Code. 17882035
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17882035 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17882035 Supplier Invitrogen™ Supplier No. 900T00
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

This kit contains the key components required for the analysis of 10 Sandwich ELISA plates. It has been specifically formulated to be used in conjunction with PeproTech’s Standard TMB ELISA Development Kits.

  • Contains the key components required for the analysis of ten sandwich ELISA plates
  • Specifically formulated to be used in conjunction with PeproTech standard TMB ELISA development kits.

Included solutions and materials:

  • PBS
  • Wash buffer
  • Blocking buffer
  • Diluent
  • TMB liquid substrate
  • ELISA microplates
  • Sealing film
  • Stop solution
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Buffer
Quantity 1 Kit
Detection Method Colorimetric
For Use With (Application) ELISA
Product Line PeproTech
Substrate Type TMB, HRP
compliance-icons compliance-icons
Product Identifier
  • TMB ELISA Buffer Kit, PeproTech
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P390-Absorb spillage to prevent material damage.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.