missing translation for 'onlineSavingsMsg'
Learn More

Abcam TMB ELISA Substrate (High Sensitivity), 100 mL

Product Code. 32115479 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115479 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115479 Supplier Abcam Supplier No. ab171523100mL
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Abcam TMB ELISA Substrate (High Sensitivity).

  • Species Reactivity: Species independent
  • Storage: Store in the dark

Specifications

Product Type ELISA Substrate
Quantity 100 mL
For Use With (Application) ELISA, Immunohistochemistry (IHC)
Content And Storage Store in the dark
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.