missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress TMB monosulfate
Specifications
Specifications
| Quantity | 100 mg |
| Storage Requirements | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Solubility | DMSO : ≥ 28 mg/mL (82.74 mM) ∣ H2O : 2.78 mg/mL (8.21 mM; ultrasonic and warming and heat to 60°C) |
| Shipping Condition | Room temperature |
| CAS | 54827-18-8 |
| Molecular Weight (g/mol) | 338.42 g/mol |
| Synonym | BM blue monosulfate ∣ Sure Blue TMB monosulfate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?