missing translation for 'onlineSavingsMsg'
Learn More

TMPyP4, TMPyP4, TRC

Product Code. 30134433
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134433 50 mg 50mg
30132490 100 mg 100mg
30008521 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134433 Supplier Toronto Research Chemicals Supplier No. TRCT46001050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C72H66N8O12S4
Molecular Weight (g/mol) 1363.6
Synonym TMP 1363, TMPyP 4 4-Methylbenzenesulfonate, TMPyP 4 Tosylate, TMPyP4 Tetra(p-toluene Sulfonate), TPPT 4, 5,10,15,20-Tetrakis(N-methylpyridinium-4-yl)-21H,23H-porphyrin tetrakis(p-toluenesulfonate), 5,10,15,20-Tetrakis(1-methylpyridinium-4-yl)porphyrin Tetratosylate, 4,4',4' ',4' ' '-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakis[1-methyl-pyridinium salt with 4-Methylbenzenesulfonic Acid
IUPAC Name 4-methylbenzenesulfonate;(5Z,10Z,14Z,19Z)-5,10,15,20-tetrakis(1-methylpyridin-1-ium-4-yl)-21,23-dihydroporphyrin
SMILES Cc1ccc(cc1)S(=O)(=O)[O-].Cc2ccc(cc2)S(=O)(=O)[O-].Cc3ccc(cc3)S(=O)(=O)[O-].Cc4ccc(cc4)S(=O)(=O)[O-].C[n+]5ccc(cc5)c6c7C=Cc([nH]7)c(c8cc[n+](C)cc8)c9ccc(n9)c(c%10cc[n+](C)cc%10)c%11ccc([nH]%11)c(c%12cc[n+](C)cc%12)c%13ccc6n%13

Specifications

CAS 36951-72-1
Quantity 50 mg
InChI Formula InChI=1S/C44H37N8.4C7H8O3S/c1-49-21-13-29(14-22-49)41-33-5-7-35(45-33)42(30-15-23-50(2)24-16-30)37-9-11-39(47-37)44(32-19-27-52(4)28-20-32)40-12-10-38(48-40)43(36-8-6-34(41)46-36)31-17-25-51(3)26-18-31;4*1-6-2-4-7(5-3-6)11(8,9)10/h5-28H,1-4H3,(H,45,46,47,48);4*2-5H,1H3,(H,8,9,10)/q+3;;;;/p-3/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;;;
Formula Weight 1362.3683
Chemical Name or Material TMPyP4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.