missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTpro™ 10plex Isobaric Label Reagent Set (Unit Mass Reporter), 1 x 5 mg

Product Code. 30245022 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245022 1 x 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245022 Supplier Thermo Scientific™ Supplier No. A40000928
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ TMTpro 10plex Label Reagent Set (Unit Mass Reporter), 1 x 5 mg enables multiplexing of up to 10 samples for simultaneous analysis on lower resolution MS instruments.

The Thermo Scientific™ TMTpro 10plex Label Reagent Set (Unit Mass Reporter), 1 x 5 mg enables multiplexing of up to 10 samples for simultaneous analysis on lower resolution MS instruments.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type 32plex Label Reagent Matched Set
Product Line TMTpro
Starting Material Peptides
Final Product Type peptide
Detection Method Mass Spectrometry
Workflow Step Peptide labeling
Quantity 1 x 5 mg
Form Powder

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.