missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTpro™ 16plex Label Reagent Set

Product Code. 17961691 Shop All Gibco Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 x 0.5 mg (per tag)
1 x 5 mg (per tag)
6 x 0.5 mg (per tag)
6 x 50 μg, 96 reactions
Förpackningsstorlek:
5mg
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Förpackningsstorlek
17961691 6 x 50 μg, 96 reactions Each
16365528 1 x 5 mg (per tag) 5mg
16426210 1 x 0.5 mg (per tag) Each
16498580 6 x 0.5 mg (per tag) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Regulatory: A license is required to purchase this product; ROYAL ORDER OF APRIL 12, 1974.
Produktkode 17961691 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' A58334
Black Friday Starts Here
Use promo code 27984
Se alle kampagner
 Restricted Item
In Stock
Tilføj til indkøbskurv
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik
Regulatory: A license is required to purchase this product; ROYAL ORDER OF APRIL 12, 1974.

The Thermo Scientific TMTpro 16plex Label Reagent Set enables multiplexing of up to 16 samples for multiplexed protein identification and quantitation using tandem mass spectrometry (MS/MS).

TMTpro label reagents are the next generation of tandem mass tags, designed to increase multiplexing without compromising protein identification and quantitation. The structures of the TMTpro and TMT tags are similar in being isobaric and amine reactive, but the TMTpro tag has a longer spacer region and isobutyl proline mass reporter. After fragmentation, each TMTpro tag generates a unique reporter mass (i.e., TMTpro 126-134Da) in the low-mass region of the high-resolution MS/MS spectrum used for multiplexed relative quantitation of protein expression.

The advantages of TMTpro tags include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. With these capabilities, the TMTpro 16plex label reagents are suited for applications requiring higher multiplexing and fewer missing values, such as inhibitor dose response experiments, time course experiments, thermal shift assays, or single cell analysis. When combined with the industry leading, high-resolution Orbitrap instruments and software, TMTpro reagents provide integrated total solutions for quantitative protein expression analysis.

Features of TMTpro 16plex Label Reagent Sets include:
• Multiplex—concurrent MS analysis of up to 16 samples derived from cells, tissues, or biological fluids
• Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
• Efficient—amine-reactive, NHS ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
• Compatible—optimized for use with high resolution Thermo Scientific Orbitrap MS/MS platforms, such as the Q Exactive and Orbitrap Fusion Tribrid instrument series, including the Orbitrap Eclipse Tribrid and Ascend and Orbitrap Exploris 480 mass spectrometers with data analysis fully supported by Proteome Discoverer 2.3

For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES902 or ES003).

TRUSTED_SUSTAINABILITY

Tekniske data

Formulation Liquid in stabilized DMSO solution
For Use With (Equipment) Mass Spectrometer
Product Type Isobaric Label Reagent Set
Product Line TMT
Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Final Product Type Peptide
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 6 x 50 μg, 96 reactions
Form Liquid
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.