missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin, MedChemExpress

Product Code. 16478306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478306 100 mg 100mg
16462136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478306 Supplier Medchem Express Supplier No. HYB0441
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad spectrum aminoglycoside antibiotic that is widely used in the treatment of moderate to severe bacterial infections due to sensitive organisms.

Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad spectrum aminoglycoside antibiotic that is widely used in the treatment of moderate to severe bacterial infections due to sensitive organisms.

Specifications

Chemical Name or Material Tobramycin
CAS 32986-56-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H37N5O9
Quantity 100 mg
Synonym Nebramycin Factor 6 Deoxykanamycin B
Solubility Information H2O : ≥ 100 mg/mL (213.90 mM) ∣DMSO : 2 mg/mL (4.28 mM; Need ultrasonic)
SMILES O[C@@H]([C@@H]1O[C@@]([C@@H](C[C@@H]2O)N)([H])O[C@@H]2CN)[C@H]([C@@H](C[C@@H]1N)N)O[C@@]([C@@H]([C@@H](N)[C@@H]3O)O)([H])O[C@@H]3CO
Molecular Weight (g/mol) 467.51
Formula Weight 467.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.