missing translation for 'onlineSavingsMsg'
Learn More

Tocofersolan, MedChemExpress

Product Code. 16438588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438588 10 g 10g
16411048 5 g 5g
16443796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16438588 Supplier Medchem Express Supplier No. HYB0717
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tocofersolan is synthetic polyethylene glycol derivative of α-tocopherol. Tocofersolan is an orally active and water-soluble analog of vitamin E. Tocofersolan can reduce neurobehavioral deficits in zebrafish embryos exposed to moderate and high concentrations of BaP during early development. Tocofersolan shows antioxidant activity. Tocofersolan can be used to provide an orally bioavailable source of vitamin E.

Tocofersolan is synthetic polyethylene glycol derivative of α-tocopherol. Tocofersolan is an orally active and water-soluble analog of vitamin E. Tocofersolan can reduce neurobehavioral deficits in zebrafish embryos exposed to moderate and high concentrations of BaP during early development. Tocofersolan shows antioxidant activity. Tocofersolan can be used to provide an orally bioavailable source of vitamin E.

Specifications

Chemical Name or Material Tocofersolan
CAS 9002-96-4
Color Off-White
Purity Grade Notes Research
Quantity 10 g
Synonym TPGS D-α-Tocopherol polyethylene glycol 1000 succinate Vitamin E-TPGS
Solubility Information H2O : 100 mg/mL (66.09 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (66.09 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (33.05 mM; Need ultrasonic)
SMILES CC1=C(CCC(CCCC(C)CCCC(C)CCCC(C)C)(C)O2)C2=C(C)C(C)=C1OC(CCC(OCCO)=O)=O.[n]
Molecular Weight (g/mol) 1513.0
Formula Weight 1513.0
Percent Purity 96.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.