Learn More
Tocris Bioscience™ [D-Lys3]-GHRP-6
Product Code. 18728826
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 18728826
Brand: Tocris Bioscience™ 1922/5
Ghrelin receptor antagonist
[D-Lys3]-GHRP-6 is an antagonist at the ghrelin receptor (GHS-R1a) (IC50 = 0.9 μM). Also weakly binds to melanocortin receptors (Ki = 26-120 μM). Centrally active in vivo.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mg