Learn More
Tocris Bioscience™ FSLLRY-NH2
GREENER_CHOICE
Product Code. 18736857
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
18736857 1 mg 1mg
1 options
Product Code. 18736857 Supplier Tocris Bioscience™ Supplier No. 4751/1
PAR2 peptide antagonist
FSLLRY-NH2 is a selective PAR2 peptide antagonist. Reverses taxol -induced mechanical allodynia, heat hyperalgesia and PKC activation in ICR mice. Blocks ERK activation and collagen production in isolated cardiac fibroblasts. Also reduces symptoms in a mouse model of dermatophyte-associated itch.
TRUSTED_SUSTAINABILITY
Specifications
Purity 0.95
Quantity 1 mg