missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Hexa-D-arginine
Furin inhibitor
Brand: Tocris Bioscience™ 4711/1
This item is not returnable.
View return policy
Description
Hexa-D-arginine is an inhibitor of furin (Ki values are 0.106, 0.58 and 13.2 μM for furin, PACE4 and PC1 respectively).Specifications
| 5045 | |
| Store at -20°C | |
| M.W. (Observed): 954.16 |
| 95% | |
| Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
| 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?