missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tolazoline hydrochloride, 99%

Product Code. 10011540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10011540 50 g Plastic bottle 50g
10370954 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011540 Supplier Thermo Scientific Chemicals Supplier No. 138980500
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59-97-2 | C10H12N2·HCl | 196.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-97-2
Molecular Formula C10H12N2·HCl
Molecular Weight (g/mol) 196.72
MDL Number MFCD00012693
InChI Key RHTNTTODYGNRSP-UHFFFAOYSA-N
Synonym tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride
PubChem CID 6048
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC=C2.Cl

Specifications

Melting Point 172.0°C to 176.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 9666
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in chloroform, freely soluble in alcohol, slightly soluble in ether and ethyl acetate
Formula Weight 196.72
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tolazoline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Tolazoline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.