missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tolcapone

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Tolcapone is a COMT inhibitor. Inhibits both brain and peripheral COMT. Also binds transthyretin (TTR) with high affinity (Kd1 and Kd2 values are 21 and 58 nM, respectively). Inhibits TTR aggregation in human plasma and prevents TTR-induced cytotoxicity in vitro. Stabilizes TTR in mice and humans in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone |
| CAS | 134308-13-7 |
| Synonym | Tasmar, Ro 40-7592 |
| Quantity | 10 mg |
| Target | Catechol O-Methyltransferase (COMT) Inhibitors |
| Molecular Formula | C14H11NO5 |
| Formula Weight | 273.24 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction