missing translation for 'onlineSavingsMsg'
Learn More

Tolmetin sodium salt dihydrate, 98+%

Product Code. 15494989
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494989 25 g 25g
15474989 1 g 1g
15484989 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494989 Supplier Thermo Scientific Alfa Aesar Supplier No. J62663.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An NSAID compound | CAS: 64490-92-2 | C15H18NNaO5 | 315.301 g/mol

Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 μM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64490-92-2
Molecular Formula C15H18NNaO5
Molecular Weight (g/mol) 315.301
MDL Number MFCD00150761
InChI Key QQILXENAYPUNEA-UHFFFAOYSA-M
Synonym tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
PubChem CID 23677829
ChEBI CHEBI:72014
IUPAC Name sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]

Specifications

Color White
Quantity 25 g
Merck Index 14,9518
Solubility Information Soluble in water
Formula Weight 315.30 (279.27 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tolmetin sodium salt dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.