missing translation for 'onlineSavingsMsg'
Learn More

Solstice Toluene

Product Code. 16665803
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16665803 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 16665803 Supplier Solstice Supplier No. 32249S18LRC
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, phenylmethane
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

pH No data available
Melting Point -95°C
Color Colorless
CAS Max % 100.00%
Density 0.867 g/cm3 (20°C)
Quantity 18 L
Flash Point 4°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Vapor Pressure 38 hPa (25°C)
Viscosity No data available
UN Number UN1294
Beilstein 635760
Concentration 100.00%
Boiling Point 110.6°C
Percent Purity ≥99.70%
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.