missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15646690
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
1 L
4 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15646690 1 L Glass Bottle 1 L
15698440 4 L Aluminum Bottle 4 L
15656690 4 x 4 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15646690 Supplier Honeywell-Riedel-de Haen Supplier No. 6767561L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -93°C (literature)
CAS Max % 100.0000%
Density 0.865 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 4°C
Packaging Glass Bottle
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110.6°C
Formula Weight 92.14
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Toluene, ACS Reagent, ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.