missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS, 99.5% min

Product Code. 11358277
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358277 1 L 1 L
11490520 500 mL 500mL
11338277 4 x 1 L 4 L
11348277 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11358277 Supplier Thermo Scientific Alfa Aesar Supplier No. 031755.K2
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 108-88-3 | C7H8 | 92.14 g/mol

Toluene is used as solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Color Colorless
Appearance Colorless liquid
Density 0.867
Quantity 1 L
Flash Point 4°C (40°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range 99.5%
Vapor Pressure 29 hPa (22mm Hg) at 20°C
Beilstein 635760
Boiling Point 111°C
Solubility Information Very slightly soluble in water. Miscible with alcohol,chloroform,ether,glacial acetic acid,carbon disulfideMiscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene, ACS
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: toluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.