missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS Reagent Grade, Ricca Chemical

Product Code. 31751068
Change view
Click to view available options
Quantity:
4 L
20 L
Unit Size:
20 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31751068 20 L 20 L
31750890 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 31751068 Supplier Ricca Chemical Company Supplier No. RSOT020020E4
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

CAS Max % 100
Quantity 20 L
Packaging Metal Can
Residue after Evaporation max. 0.001%
Water max. 0.03%
Boiling Point 111°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.