missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, For pesticide residue analysis according to FDA, Honeywell Riedel-de Haën™
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| Color | Colorless |
| Density | 0.865g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.07 |
| Flash Point | 4°C (39.2°F) |
| Ignition Point | 997°F |
| Linear Formula | C6H5CH3 |
| Vapor Pressure | 22 mmHg (20°C), 26 mmHg (25°C) |
| UN Number | UN1294 |
| Beilstein | 635760 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: toluene
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?