missing translation for 'onlineSavingsMsg'
Learn More

Toluene-d8, for NMR, 99.8 atom % D

Product Code. 16358216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16358216 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16358216 Supplier Thermo Scientific Acros Supplier No. 464240100
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    146.00 € / 10mL
    Save 5.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.9400 g/mL
Flash Point 4°C
Refractive Index 1.4940
Specific Gravity 0.94
Quantity 10 mL
CAS 2037-26-5
Melting Point -84.0°C
Boiling Point 110.0°C
Molecular Formula C7H8
Linear Formula C6D5CD3
Solubility Information Solubility aq. soln.: almost insoluble (0.5 g/L (at 20°C).)
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
IUPAC Name 1-(²H₃)methyl(²H₅)benzene
Molecular Weight (g/mol) 100.19
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene-d8
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Toluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.