missing translation for 'onlineSavingsMsg'
Learn More

Toluene-d8, for NMR, 99+ atom % D

Product Code. 10114230 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10114230 10 mL Glass Bottle 10mL
10275590 25 mL Glass Bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10114230 Supplier Thermo Scientific Supplier No. 166390100
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    110.00 € / 10mL
    Save 11.00 €
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
MDL Number MFCD00044638
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
Synonym toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
PubChem CID 74861
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
SMILES CC1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Density 0.9400g/mL
Deuterium (D) 99.0% min.
Flash Point 4°C
Packaging Glass Bottle
Refractive Index 1.494
Quantity 10 mL
Melting Point -84.0°C
Boiling Point 110.0°C
Linear Formula C6D5CD3
Beilstein 05, IV, 797
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8, 99+ atom % D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene-d8
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.