missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Honeywell Riedel-de Haën™

Product Code. 15603750
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15603750 2.5 L 2.5 L
15693740 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15603750 Supplier Honeywell Riedel-de Haën Supplier No. 322492.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Density 0.865g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Linear Formula C6H5CH3
Vapor Pressure 22mmHg (20°C), 26mmHg (25°C)
UN Number 1294
Beilstein 635760
Formula Weight 92.14g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Toluene, Reag. Ph. Eur. reag. ISO
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.