missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Tomoxetine hydrochloride

Product Code. 18045982
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045982 50 mg 50mg
18005522 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18045982 Supplier R&D Systems™ Supplier No. 2011/50
  • ONLINE EXCLUSIVE PRICE
    1156.11 €
    1034.25 € / 50mg
    Save 121.86 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective noradrenalin re-uptake inhibitor

Tomoxetine hydrochloride is a potent and selective noradrenalin re-uptake inhibitor (Ki values are 5, 77 and 1451 nM for inhibition of radioligand binding to human NET, SERT and DAT respectively). Displays minimal affinity for a range of other neurotransmitter receptors and transporters (Ki > 1 μM). Antidepressant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82248-59-7
Molecular Formula C17H22ClNO

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 291.82
Purity >99%
Molecular Weight (g/mol) 291.819
Chemical Name or Material Tomoxetine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.