missing translation for 'onlineSavingsMsg'
Learn More

Topiramate, TRC

Product Code. 30010448
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010448 50 mg 50mg
17981655 1 g 1g
17953775 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010448 Supplier Toronto Research Chemicals Supplier No. TRCT54025050MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97240-79-4
Molecular Formula C12 H21 N O8 S
Molecular Weight (g/mol) 339.36
Synonym Topiramate, 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran, β-D-fructopyranose deriv., 2,3:4,5-Bis-O-(1-methylethylidene) β-D-fructopyranose 1-sulfamate, 2,3:4,5-Bis-O-(1-methylethylidene) β-D-fructopyranose sulfamate, Epitoma, Epitomax, McN 4853, RWJ 17021, TPM, Topamac, Topamax, Topimax, Topina, Topomax
IUPAC Name [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[5,4-b:5',3'-d]pyran-3a-yl]methyl sulfamate
SMILES CC1(C)O[C@@H]2CO[C@@]3(COS(=O)(=O)N)OC(C)(C)O[C@H]3[C@@H]2O1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1
Formula Weight 339.0988
Percent Purity >95%
Chemical Name or Material Topiramate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.