missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, without competent cells

Product Code. 11503837
Change view
Click to view available options
Quantity:
10 Reactions
25 Reactions
Unit Size:
25 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11503837 25 Reactions 25 reactions
15751927 10 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11503837 Supplier Invitrogen™ Supplier No. 450640
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water

Includes the pCR™II-TOPO TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable cloning method, featuring a 5-minute cloning reaction and 3-step protocol, and yielding up to 95% recombinants.

Includes the pCR™II-TOPO TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable cloning method, featuring a 5-minute cloning reaction and 3-step protocol, and yielding up to 95% recombinants. Resistant to Kanamycin (KanR) andZeocin™ (ZeoR)
Kit Options: TOPO TA Cloning Dual Promoter Kits
TOPO TA Cloning Dual Promoter kits can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

  • General cloning: TOP10 cells (Cat. Nos. K4600-J10, K4600-01, K4600-40)
  • High-efficiency cloning: TOP10 Electrocomp™ cells (Cat. Nos. K4660-01, K4660-40)
  • General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. Nos. K4620-01, K4620-40)
  • Fast growth: Mach1™ -T1R chemically competent E. coli (Cat. No. K4610-20)
  • Repressor/induction needs: TOP10F' cells (Cat. Nos. K4650-01, K4650-40)
  • Provide your own cells (Cat. Nos. 450640 and 452640)

  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and SP6 promoters for efficient in vitro transcription
  • M13 forward and reverse primer sites for sequencing
  • 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
  • Kanamycin and ampicillin resistance for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants

Cloning, PCR Cloning

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR), Kanamycin (KanR)
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.
Format Kit
Product Line TOPO
Product Type Cloning Kit
Promoter T7, SP6
Quantity 25 Reactions
Shipping Condition Dry Ice
Vector TOPO-TA Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.