missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Topotecan hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Topotecan hydrochloride is a DNA topoisomerase I inhibitor; induces rapid apoptotic death in human B-lineage acute lymphoblastic leukemia (ALL) cells. Improves survival in three mouse models of poor prognosis ALL; downregulates B-Myb and MycN expression in neuroblastoma cells. Water-soluble analog of Camptothecin (Cat. No. 1100). Orally bioavailable. Topotecan disrupts the interaction between NHP2L1 and U4 RNA by binding to U4 and inhibits RNA splicing.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 457.91 |
| Percent Purity | >98% |
| Chemical Name or Material | Topotecan hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction