missing translation for 'onlineSavingsMsg'
Learn More

Topotecan Hydrochloride, TRC

Product Code. 30456308
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456308 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456308 Supplier Toronto Research Chemicals Supplier No. TRCT542500100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Topotecan Hydrochloride
CAS 119413-54-6
Molecular Formula C23 H23 N3 O5 . Cl H
Quantity 100 mg
Synonym (4S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione Hydrochloride (1:1), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione','10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-','monohydrochloride','(4S)- (9CI), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione','10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-','monohydrochloride','(S)-, (S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[l,2-b]quinoline-3,14(4H,12H)-dione monohydrochloride, Hycamtin, NSC 609669, Nogitecan hydrochloride, SKF 104864A, SKFS 104864A, Topotecan chlorohydrate, Topotecan hydrochloride, Xinze
InChI Formula InChI=1S/C23H23N3O5.ClH/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20;/h5-8,27,30H,4,9-11H2,1-3H3;1H/t23-;/m0./s1
SMILES Cl.CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CN(C)C)c(O)ccc5nc34)C2=O
IUPAC Name (19S)-8-[(dimethylamino)methyl]-19-ethyl-7,19-dihydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2,4(9),5,7,10,15(20)-heptaene-14,18-dione;hydrochloride
Molecular Weight (g/mol) 457.91
Formula Weight 457.1404
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.