missing translation for 'onlineSavingsMsg'
Learn More

Toray Carbon Paper, TGP-H-60, 19x19cm

Product Code. 15402227
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Cardboard Box
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15402227 1 Ea. Cardboard Box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15402227 Supplier Thermo Scientific Alfa Aesar Supplier No. 045356.KS
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-42-5 | C | 12.01 g/mol

Toray Carbon Paper, TGP-H-60 is used as a gas diffusion layer in fuel cell applications. It is suitable for use as a catalyst backing layer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7782-42-5
Packaging Cardboard Box
Molecular Formula C
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
Quantity 1 Ea.
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
Solubility Information Insoluble in water.
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.01
PubChem CID 5462310
ChEBI CHEBI:27594
Chemical Name or Material Toray Carbon Paper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.