Learn More
3M™ Torque™ Corded Earplugs
Product Code. 15789777
Change view
Click to view available options
Color: Gray
Product Code.
15789777 Color: Gray
1 options
Product Code. 15789777 Supplier 3M™ Supplier No. 7000052852
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Torque™ Corded Earplugs perform as tough as they look by providing protection for noisy environments.
  • Earplugs perform as tough as they look by providing excellent protection for noisy environments
  • The screw threads are molded into the foam for more than looks, twisting the plug while pushing into the ear canal can achieve an effective seal
  • Earplugs are reusable, made from ultra-soft foam that provides all day long comfort
  • Insertion stem helps eliminate the need to touch the tip when fitting
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Pairs per Package 120 Pairs/Pk.
Corded Corded
Product Type Earplugs