missing translation for 'onlineSavingsMsg'
Learn More

Tosylmethyl Isocyanide, TRC

Product Code. 30451445
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451445 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451445 Supplier Toronto Research Chemicals Supplier No. TRCT66750050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tosylmethyl Isocyanide
CAS 36635-61-7
Molecular Formula C9H9NO2S
Quantity 50 g
Synonym 1-[(Isocyanomethyl)sulfonyl]-4-methylbenzene, (4-Methylphenylsulfonyl)methyl Isocyanide, (p-Tolylsulfonyl)methyl isocyanide, 4-Toluenesulfonylmethyl Isocyanide, 4-Tolylsulfonylmethyl Isocyanide, Isocyanomethyl p-Tolyl Sulfone, NSC 631633, TOSMIC, p-Toluenesulfonylmethyl Isocyanide, p-Toluenesulfonylmethylisonitrile, p-Tosylmethyl Isocyanide, p-Tosylmethyl Isonitrile
InChI Formula InChI=1S/C9H9NO2S/c1-8-3-5-9(6-4-8)13(11,12)7-10-2/h3-6H,7H2,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)C[N+]#[C-]
IUPAC Name 1-(isocyanomethylsulfonyl)-4-methylbenzene
Molecular Weight (g/mol) 195.24
Formula Weight 195.0354
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.